Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CCDC78 SIRNA Assays ›
Gene Symbol:
CCDC78
Gene Name:
coiled-coil domain containing 78
Gene Aliases:
C16orf25, CNM4, JFP10, hsCCDC78
Chromosome Location:
Chr.16: 722578 - 726880 on Build GRCh38
UniGene ID:
Hs.381943
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
215902
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CCDC78
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001031737.2
NP_001026907.2
14
1418
XM_006720838.1
XP_006720901.1
12
1699
XM_006720843.3
XP_006720906.1
13
1856
XM_011522356.1
XP_011520658.1
9
1924
XM_011522357.1
XP_011520659.1
11
1912
XM_011522358.2
XP_011520660.1
9
1900
XM_011522359.1
XP_011520661.1
9
1891
XM_011522360.1
XP_011520662.1
11
1879
XM_011522361.1
XP_011520663.1
10
1852
XM_011522362.1
XP_011520664.1
9
1980
XM_011522363.1
XP_011520665.1
10
1848
XM_011522364.1
XP_011520666.1
10
1824
XM_011522365.1
XP_011520667.1
10
1711
XM_011522366.1
XP_011520668.1
10
1689
XM_011522367.1
XP_011520669.1
9
1543
XM_011522368.1
XP_011520670.1
11
1531
XM_011522369.1
XP_011520671.1
11
1489
XM_011522370.1
XP_011520672.1
10
1308
XM_011522371.2
XP_011520673.1
10
1300
XM_017022929.1
XP_016878418.1
10
1828
XM_017022930.1
XP_016878419.1
13
1594
XM_017022931.1
XP_016878420.1
3
1105
XM_017022932.1
XP_016878421.1
6
946
GenBank mRNA
AK057178.1
-
13
1660
AK091831.1
-
10
1885
AK127495.1
-
4
2862
AK128538.1
-
4
3267
AK128575.1
-
5
3061
BC042110.1
-
14
1404
5177109
Back To Top
More Information
Gene Ontology Categories:
Function(s)
skeletal muscle contraction
cell projection organization
de novo centriole assembly
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-x5whk:80/10.239.163.84:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0