Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: B9D1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243473.2 | NP_001230402.1 | 5 | 457 | ||
NM_001243475.2 | NP_001230404.1 | 5 | 653 | |||
NM_001321214.1 | NP_001308143.1 | 5 | 550 | |||
NM_001321215.1 | NP_001308144.1 | 5 | 550 | |||
NM_001321216.1 | NP_001308145.1 | 5 | 550 | |||
NM_001321217.1 | NP_001308146.1 | 5 | 550 | |||
NM_001321218.1 | NP_001308147.1 | 5 | 550 | |||
NM_001321219.1 | NP_001308148.1 | 5 | 550 | |||
NM_015681.4 | NP_056496.1 | 5 | 550 | |||
XM_005256607.2 | XP_005256664.1 | 5 | 522 | |||
XM_005256610.1 | XP_005256667.1 | 5 | 525 | |||
XM_017024452.1 | XP_016879941.1 | 8 | 2914 | |||
GenBank mRNA | AB030506.1 | - | 5 | 539 | ||
AK293815.1 | - | 5 | 523 | |||
AK297782.1 | - | 5 | 440 | |||
AK309320.1 | - | 5 | 642 | |||
BC002944.2 | - | 5 | 481 | 3953287 | ||
BI833202.1 | - | 5 | 545 |