Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSME1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281528.1 | NP_001268457.1 | 4 | 255 | ||
NM_001281529.1 | NP_001268458.1 | 4 | 255 | |||
NM_006263.3 | NP_006254.1 | 4 | 255 | |||
NM_176783.2 | NP_788955.1 | 4 | 255 | |||
GenBank mRNA | AK311490.1 | - | 2 | 456 | ||
AK312211.1 | - | 4 | 190 | |||
BC000352.2 | - | 4 | 241 | 2961448 | ||
BC007503.2 | - | 4 | 241 | 2961448 | ||
BJ993879.1 | - | 4 | 255 | |||
BM470561.1 | - | 4 | 191 | |||
BQ060125.1 | - | 4 | 213 | |||
BT019337.1 | - | 4 | 150 | |||
BU192084.1 | - | 4 | 186 | |||
CR456780.1 | - | 4 | 150 | |||
CR541658.1 | - | 4 | 150 | |||
DQ185047.1 | - | 2 | 86 | |||
GQ472237.1 | - | 4 | 241 | |||
L07633.1 | - | 4 | 242 |