Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNRNP70 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301069.1 | NP_001287998.1 | 10 | 1712 | ||
NM_003089.5 | NP_003080.2 | 10 | 1739 | |||
XM_005259178.1 | XP_005259235.1 | 4 | 772 | |||
XM_011527240.2 | XP_011525542.2 | 5 | 958 | |||
XM_011527241.2 | XP_011525543.1 | 4 | 925 | |||
GenBank mRNA | AK027127.1 | - | 3 | 1265 | ||
AK056808.1 | - | 3 | 2829 | |||
AK096783.1 | - | 11 | 2902 | |||
AK293120.1 | - | 10 | 1458 | |||
AL117399.1 | - | 2 | 2731 | |||
AL117507.1 | - | 3 | 1771 | |||
AL832819.1 | - | 3 | 2980 | |||
BC000342.2 | - | 2 | 633 | |||
BC001315.1 | - | 10 | 1412 | 3454479 | ||
M22636.1 | - | 10 | 1403 | |||
X06811.1 | - | 1 | 147 | |||
X06814.1 | - | 3 | 679 | |||
X06815.1 | - | 11 | 1428 | |||
X06816.1 | - | 1 | 274 | |||
X07401.1 | - | 4 | 778 | |||
X07402.1 | - | 1 | 157 | |||
X07403.1 | - | 1 | 107 | |||
X84841.1 | - | 9 | 1232 |