Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COMMD7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099339.1 | NP_001092809.1 | 4 | 901 | ||
NM_053041.2 | NP_444269.2 | 4 | 904 | |||
XM_005260299.3 | XP_005260356.1 | 4 | 891 | |||
XM_005260300.3 | XP_005260357.1 | 4 | 888 | |||
XM_011528604.2 | XP_011526906.1 | 4 | 891 | |||
XM_017027685.1 | XP_016883174.1 | 4 | 888 | |||
XM_017027686.1 | XP_016883175.1 | 4 | 891 | |||
XM_017027687.1 | XP_016883176.1 | 4 | 891 | |||
GenBank mRNA | AK027893.1 | - | 4 | 351 | ||
AK096307.1 | - | 4 | 893 | |||
AK097059.1 | - | 5 | 428 | |||
AY542162.1 | - | 4 | 287 | |||
BC000628.2 | - | 4 | 337 | 3343149 | ||
BC047440.1 | - | 4 | 465 | 5275167 |