Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COMMD7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099339.1 | NP_001092809.1 | 9 | 1867 | ||
NM_053041.2 | NP_444269.2 | 9 | 1870 | |||
XM_011528604.2 | XP_011526906.1 | 9 | 2321 | |||
XM_017027685.1 | XP_016883174.1 | 9 | 2318 | |||
XM_017027686.1 | XP_016883175.1 | 7 | 1716 | |||
GenBank mRNA | AK027893.1 | - | 9 | 1317 | ||
AK096307.1 | - | 9 | 1859 | |||
AK097059.1 | - | 9 | 1958 | |||
BC000628.2 | - | 9 | 1303 | 3343149 | ||
BC047440.1 | - | 9 | 1431 | 5275167 |