Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: P2RX5-TAX1BP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037928.1 | 13, 14 | 5222 | |||
GenBank mRNA | AB209623.1 | - | 13, 14 | 5010 |
Gene Symbol: TAX1BP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014604.3 | NP_055419.1 | 2, 3 | 320 | ||
GenBank mRNA | AF028823.2 | - | 2, 3 | 323 | ||
AF234997.1 | - | 2, 3 | 214 | |||
AF277318.1 | - | 2, 3 | 193 | |||
AK315408.1 | - | 2, 3 | 219 | |||
BC023980.1 | - | 2, 3 | 223 | 4069168 | ||
U90913.1 | - | 2, 3 | 201 |