Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPOPL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001664.2 | NP_001001664.1 | 11 | 2949 | ||
XM_005263655.4 | XP_005263712.1 | 12 | 2912 | |||
XM_005263656.4 | XP_005263713.1 | 12 | 2841 | |||
XM_005263657.4 | XP_005263714.1 | 11 | 2754 | |||
GenBank mRNA | BC071613.1 | - | 11 | 2927 | 5275316 | |
BX538143.1 | - | 2 | 1568 |