Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C4orf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001701.3 | NP_001001701.2 | 2 | 671 | ||
NM_001170330.1 | NP_001163801.1 | 3 | 792 | |||
GenBank mRNA | AF075061.1 | - | 2 | 479 | ||
AK025703.1 | - | 2 | 435 | |||
BC017399.1 | - | 2 | 494 | 4108498 | ||
CN361054.1 | - | 2 | 222 |