Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RTP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004312.2 | NP_001004312.2 | 1 | 454 | ||
XM_017006301.1 | XP_016861790.1 | 4 | 648 | |||
XM_017006302.1 | XP_016861791.1 | 4 | 599 | |||
GenBank mRNA | AY562236.1 | - | 1 | 25 | ||
BC068081.1 | - | 1 | 454 | 6212901 |