Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OR10X1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004477.1 | NP_001004477.1 | 1 | 268 |
Set Membership: |
G-protein coupled receptor |