Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRC4C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001258419.1 | NP_001245348.1 | 7 | 2117 | ||
NM_020929.2 | NP_065980.1 | 5 | 1948 | |||
NR_047673.1 | 3 | 2449 | ||||
NR_047674.1 | 2 | 2396 | ||||
XM_011520238.2 | XP_011518540.1 | 8 | 2905 | |||
XM_011520239.2 | XP_011518541.1 | 9 | 2971 | |||
XM_011520240.2 | XP_011518542.1 | 7 | 2825 | |||
XM_011520241.2 | XP_011518543.1 | 4 | 3540 | |||
XM_011520242.2 | XP_011518544.1 | 4 | 3526 | |||
XM_011520243.2 | XP_011518545.1 | 7 | 2852 | |||
XM_011520244.2 | XP_011518546.1 | 6 | 2772 | |||
XM_017018070.1 | XP_016873559.1 | 8 | 2901 | |||
XM_017018071.1 | XP_016873560.1 | 8 | 2891 | |||
XM_017018072.1 | XP_016873561.1 | 8 | 2754 | |||
XM_017018073.1 | XP_016873562.1 | 6 | 2613 | |||
XM_017018074.1 | XP_016873563.1 | 4 | 3521 | |||
XM_017018075.1 | XP_016873564.1 | 5 | 2560 | |||
XM_017018076.1 | XP_016873565.1 | 3 | 3487 | |||
XM_017018077.1 | XP_016873566.1 | 6 | 2640 | |||
XM_017018078.1 | XP_016873567.1 | 5 | 3606 | |||
XM_017018079.1 | XP_016873568.1 | 6 | 2768 | |||
GenBank mRNA | AB046800.1 | - | 2 | 3381 | ||
AK289646.1 | - | 3 | 1584 | |||
AY358293.1 | - | 2 | 2227 | |||
BC041374.2 | - | 3 | 2449 | 5274422 | ||
DQ084201.1 | - | 7 | 2066 | |||
DQ084202.1 | - | 5 | 2085 |