Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OR2J3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005216.3 | NP_001005216.2 | 1 | 105 | ||
GenBank mRNA | BC136973.1 | - | 1 | 105 | 9020965 | |
BC136975.1 | - | 1 | 106 | 9020967 |