Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNASE9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001673.3 | NP_001001673.2 | 2 | 534 | ||
NM_001110356.1 | NP_001103826.1 | 3 | 586 | |||
NM_001110357.1 | NP_001103827.1 | 3 | 569 | |||
NM_001110358.1 | NP_001103828.1 | 4 | 622 | |||
NM_001110359.1 | NP_001103829.1 | 5 | 732 | |||
NM_001110360.1 | NP_001103830.1 | 4 | 697 | |||
NM_001110361.1 | NP_001103831.1 | 5 | 657 | |||
NM_001289110.1 | NP_001276039.1 | 4 | 621 | |||
GenBank mRNA | AF382949.1 | - | 1 | 462 | ||
AY665804.1 | - | 1 | 462 | |||
AY907670.1 | - | 2 | 534 | |||
BC130311.1 | - | 1 | 547 | 40146342 | ||
BC130313.1 | - | 1 | 547 | 40146344 | ||
EF061296.1 | - | 3 | 569 | |||
EF061297.1 | - | 3 | 586 | |||
EF061298.1 | - | 4 | 621 | |||
EF061299.1 | - | 4 | 622 | |||
EF061300.1 | - | 4 | 697 | |||
EF061301.1 | - | 5 | 657 | |||
EF061302.1 | - | 5 | 732 | |||
EF061303.1 | - | 3 | 2153 | |||
EF061304.1 | - | 4 | 2188 | |||
EU414264.1 | - | 1 | 462 |