Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPATCH8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002909.3 | NP_001002909.1 | 8 | 2993 | ||
NM_001304939.1 | NP_001291868.1 | 7 | 2918 | |||
NM_001304940.1 | NP_001291869.1 | 10 | 3134 | |||
NM_001304941.1 | NP_001291870.1 | 9 | 3011 | |||
NM_001304942.1 | NP_001291871.1 | 7 | 2920 | |||
NM_001304943.1 | NP_001291872.1 | 5 | 2874 | |||
XM_011524557.1 | XP_011522859.1 | 10 | 3067 | |||
XM_011524558.1 | XP_011522860.1 | 11 | 2886 | |||
XM_011524559.2 | XP_011522861.1 | 12 | 2964 | |||
XM_011524561.2 | XP_011522863.1 | 8 | 2725 | |||
XM_017024372.1 | XP_016879861.1 | 13 | 3009 | |||
XM_017024373.1 | XP_016879862.1 | 10 | 3844 | |||
XM_017024374.1 | XP_016879863.1 | 12 | 2927 | |||
XM_017024375.1 | XP_016879864.1 | 9 | 3022 | |||
XM_017024376.1 | XP_016879865.1 | 10 | 2882 | |||
XM_017024377.1 | XP_016879866.1 | 9 | 2859 | |||
XM_017024378.1 | XP_016879867.1 | 10 | 3040 | |||
XM_017024379.1 | XP_016879868.1 | 11 | 2922 | |||
XM_017024380.1 | XP_016879869.1 | 10 | 2860 | |||
GenBank mRNA | AB011125.2 | - | 7 | 4083 | ||
BC136629.1 | - | 9 | 2810 | 9020619 | ||
BC144323.1 | - | 9 | 2810 |