Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPATCH8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002909.3 | NP_001002909.1 | 8 | 5299 | ||
NM_001304939.1 | NP_001291868.1 | 7 | 5224 | |||
NM_001304940.1 | NP_001291869.1 | 10 | 5440 | |||
NM_001304941.1 | NP_001291870.1 | 9 | 5317 | |||
NM_001304942.1 | NP_001291871.1 | 7 | 5226 | |||
NM_001304943.1 | NP_001291872.1 | 5 | 5180 | |||
XM_011524557.1 | XP_011522859.1 | 10 | 5373 | |||
XM_011524558.1 | XP_011522860.1 | 11 | 5192 | |||
XM_011524559.2 | XP_011522861.1 | 12 | 5270 | |||
XM_011524561.2 | XP_011522863.1 | 8 | 5031 | |||
XM_017024372.1 | XP_016879861.1 | 13 | 5315 | |||
XM_017024373.1 | XP_016879862.1 | 10 | 6150 | |||
XM_017024374.1 | XP_016879863.1 | 12 | 5233 | |||
XM_017024375.1 | XP_016879864.1 | 9 | 5328 | |||
XM_017024376.1 | XP_016879865.1 | 10 | 5188 | |||
XM_017024377.1 | XP_016879866.1 | 9 | 5165 | |||
XM_017024378.1 | XP_016879867.1 | 10 | 5346 | |||
XM_017024379.1 | XP_016879868.1 | 11 | 5228 | |||
XM_017024380.1 | XP_016879869.1 | 10 | 5166 | |||
GenBank mRNA | AB011125.2 | - | 7 | 6389 | ||
BC024147.2 | - | 1 | 2078 |