Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WNT11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004626.2 | NP_004617.2 | 3 | 684 | ||
XM_005274231.1 | XP_005274288.1 | 4 | 940 | |||
XM_011545238.1 | XP_011543540.1 | 4 | 970 | |||
XM_011545239.2 | XP_011543541.1 | 4 | 1178 | |||
XM_011545240.2 | XP_011543542.1 | 5 | 1134 | |||
XM_011545241.2 | XP_011543543.1 | 3 | 422 | |||
XM_017018241.1 | XP_016873730.1 | 2 | 1442 | |||
GenBank mRNA | AB070218.1 | - | 3 | 623 | ||
AK075540.1 | - | 4 | 940 | |||
AK313570.1 | - | 3 | 686 | |||
BC074790.2 | - | 3 | 603 | 30915564 | ||
BC074791.2 | - | 3 | 603 | 30915347 | ||
BC113386.1 | - | 3 | 604 | 8322438 | ||
BC113388.1 | - | 3 | 604 | 8322440 | ||
BT019492.1 | - | 3 | 561 | |||
Y12692.1 | - | 3 | 684 |