Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM87A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110503.2 | NP_001103973.1 | 1 | 218 | ||
NM_001286487.1 | NP_001273416.1 | 1 | 218 | |||
NM_015497.4 | NP_056312.2 | 1 | 218 | |||
XM_005254287.1 | XP_005254344.1 | 1 | 69 | |||
XM_011521443.2 | XP_011519745.1 | 1 | 47 | |||
XM_011521444.2 | XP_011519746.1 | 1 | 69 | |||
GenBank mRNA | AF132733.1 | - | 1 | 74 | ||
AK074870.1 | - | 1 | 67 | |||
AK075403.1 | - | 1 | 104 | |||
AK289591.1 | - | 1 | 67 | |||
AK290150.1 | - | 1 | 53 | |||
AK290484.1 | - | 1 | 62 | |||
AK309915.1 | - | 1 | 67 | |||
AK309954.1 | - | 1 | 68 | |||
BC005335.1 | - | 1 | 70 | |||
DC341748.1 | - | 1 | 191 | |||
HY036655.1 | - | 1 | 218 |