Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KRT19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002276.4 | NP_002267.2 | 1 | 387 | ||
GenBank mRNA | AK313261.1 | - | 1 | 306 | ||
BC002539.2 | - | 1 | 291 | 3138504 | ||
BC007628.1 | - | 1 | 291 | 3504206 | ||
BC010409.2 | - | 1 | 298 | 3689178 | ||
BC067744.1 | - | 1 | 306 | 30332499 | ||
BC084574.1 | - | 1 | 289 | 6730698 | ||
BG490639.1 | - | 1 | 291 | |||
BG750971.1 | - | 380 | ||||
BP328445.1 | - | 1 | 394 | |||
DA068330.1 | - | 1 | 370 | |||
Y00503.1 | - | 1 | 277 |