Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MPDZ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001261406.1 | NP_001248335.1 | 46 | 6437 | ||
NM_001261407.1 | NP_001248336.1 | 45 | 6350 | |||
NM_003829.4 | NP_003820.2 | 46 | 6449 | |||
XM_005251622.4 | XP_005251679.1 | 47 | 6485 | |||
XM_005251623.4 | XP_005251680.1 | 45 | 6275 | |||
XM_006716885.3 | XP_006716948.1 | 47 | 6534 | |||
XM_006716886.3 | XP_006716949.1 | 48 | 6693 | |||
XM_006716887.3 | XP_006716950.1 | 47 | 6637 | |||
XM_006716888.3 | XP_006716951.1 | 46 | 6398 | |||
XM_006716889.3 | XP_006716952.1 | 46 | 6389 | |||
XM_006716891.3 | XP_006716954.1 | 45 | 6302 | |||
XM_017015252.1 | XP_016870741.1 | 46 | 6401 | |||
XM_017015253.1 | XP_016870742.1 | 45 | 6287 | |||
XM_017015254.1 | XP_016870743.1 | 46 | 6486 | |||
XM_017015255.1 | XP_016870744.1 | 45 | 6334 | |||
XM_017015256.1 | XP_016870745.1 | 44 | 6188 | |||
XM_017015257.1 | XP_016870746.1 | 44 | 6175 | |||
GenBank mRNA | AB210041.1 | - | 46 | 6450 | ||
AF093419.1 | - | 46 | 6276 | |||
AJ001319.1 | - | 12 | 1463 | |||
AK058011.1 | - | 7 | 1876 | |||
AK074721.1 | - | 16 | 1938 | |||
AK091945.1 | - | 22 | 2814 | |||
AK098775.1 | - | 14 | 1714 | |||
AK294976.1 | - | 25 | 3042 | |||
AK307988.1 | - | 23 | 2854 | |||
CR936648.1 | - | 31 | 6424 | |||
CX782625.1 | - | 2 | 297 |