Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BAZ2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300905.1 | NP_001287834.1 | 10 | 2240 | ||
NM_013449.3 | NP_038477.2 | 10 | 2231 | |||
XM_011537797.2 | XP_011536099.1 | 10 | 2145 | |||
XM_011537798.2 | XP_011536100.1 | 10 | 2145 | |||
XM_011537799.2 | XP_011536101.1 | 11 | 2055 | |||
XM_011537800.2 | XP_011536102.1 | 10 | 2197 | |||
XM_011537801.2 | XP_011536103.1 | 10 | 2224 | |||
XM_011537803.2 | XP_011536105.1 | 10 | 2240 | |||
XM_011537804.2 | XP_011536106.1 | 12 | 2323 | |||
XM_011537805.2 | XP_011536107.1 | 11 | 2326 | |||
XM_011537806.2 | XP_011536108.1 | 10 | 2104 | |||
XM_011537807.2 | XP_011536109.1 | 10 | 2145 | |||
XM_011537809.2 | XP_011536111.1 | 4 | 444 | |||
XM_017018740.1 | XP_016874229.1 | 11 | 2371 | |||
XM_017018741.1 | XP_016874230.1 | 11 | 2145 | |||
XM_017018742.1 | XP_016874231.1 | 12 | 2281 | |||
GenBank mRNA | AB002312.2 | - | 9 | 2020 | ||
AF000422.1 | - | 6 | 963 | |||
AK023830.1 | - | 8 | 1866 | |||
AK023842.1 | - | 10 | 2120 | |||
CR749379.1 | - | 10 | 2232 |