Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNRNP40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004814.2 | NP_004805.2 | 8, 9 | 974 | ||
GenBank mRNA | AF090988.1 | - | 8, 9 | 943 | ||
AK074516.1 | - | 8, 9 | 957 | |||
AK123103.1 | - | 4, 5 | 1384 | |||
AK298823.1 | - | 8, 9 | 992 | |||
AK313800.1 | - | 8, 9 | 939 | |||
AL157420.1 | - | 2, 3 | 579 | |||
BC001494.2 | - | 8, 9 | 950 | 3504573 | ||
EF011621.1 | - | 8, 9 | 949 |