Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HMHB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021182.1 | NP_067005.1 | 2 | 244 | ||
GenBank mRNA | AF103884.1 | - | 2 | 244 | ||
BC096711.1 | - | 2 | 178 | |||
BC098244.1 | - | 2 | 178 | |||
BC098251.2 | - | 2 | 178 | |||
BC104412.1 | - | 2 | 178 | 40000018 | ||
BC104413.1 | - | 2 | 178 | 40000019 | ||
BC113894.1 | - | 2 | 178 | |||
BC113971.1 | - | 2 | 178 |