Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SDCCAG8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006642.3 | NP_006633.1 | 10 | 1370 | ||
XM_005273013.4 | XP_005273070.1 | 9 | 1202 | |||
XM_005273018.1 | XP_005273075.1 | 9 | 1209 | |||
XM_005273021.4 | XP_005273078.1 | 10 | 1142 | |||
XM_005273022.3 | XP_005273079.1 | 7 | 991 | |||
XM_006711727.3 | XP_006711790.1 | 10 | 1256 | |||
XM_006711728.3 | XP_006711791.1 | 9 | 1127 | |||
XM_011544021.2 | XP_011542323.1 | 11 | 1352 | |||
XM_011544022.2 | XP_011542324.1 | 11 | 1427 | |||
XM_011544023.2 | XP_011542325.1 | 11 | 1352 | |||
XM_011544024.2 | XP_011542326.1 | 11 | 1352 | |||
XM_011544026.2 | XP_011542328.1 | 11 | 1352 | |||
XM_011544028.2 | XP_011542330.1 | 9 | 1127 | |||
XM_011544030.2 | XP_011542332.1 | 3 | 1903 | |||
XM_017000103.1 | XP_016855592.1 | 11 | 1352 | |||
XM_017000104.1 | XP_016855593.1 | 9 | 1202 | |||
XM_017000105.1 | XP_016855594.1 | 10 | 1357 | |||
XM_017000106.1 | XP_016855595.1 | 10 | 1338 | |||
XM_017000107.1 | XP_016855596.1 | 11 | 1254 | |||
XM_017000108.1 | XP_016855597.1 | 13 | 1801 | |||
GenBank mRNA | AF039690.1 | - | 4 | 541 | ||
AF250731.1 | - | 10 | 1201 | |||
AK297105.1 | - | 6 | 789 | |||
AK299249.1 | - | 7 | 962 |