Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NUP58 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008564.1 | NP_001008564.1 | 10 | 1325 | ||
NM_014089.3 | NP_054808.1 | 11 | 1361 | |||
XM_005266600.3 | XP_005266657.1 | 11 | 1705 | |||
XM_011535328.1 | XP_011533630.1 | 11 | 1705 | |||
XM_011535329.2 | XP_011533631.1 | 12 | 1791 | |||
XM_017020880.1 | XP_016876369.1 | 13 | 1873 | |||
XM_017020881.1 | XP_016876370.1 | 11 | 1766 | |||
GenBank mRNA | AB007870.1 | - | 11 | 1258 | ||
AK123955.1 | - | 12 | 1195 | |||
AK302946.1 | - | 10 | 1220 | |||
AK303409.1 | - | 10 | 1129 | |||
BC001104.2 | - | 11 | 1233 | 3504343 |