Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MICA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000247.2 | NP_000238.1 | 2 | 350 | ||
NM_001177519.2 | NP_001170990.1 | 2 | 350 | |||
NM_001289152.1 | NP_001276081.1 | 2 | 1214 | |||
NM_001289153.1 | NP_001276082.1 | 2 | 1194 | |||
NM_001289154.1 | NP_001276083.1 | 2 | 1194 | |||
GenBank mRNA | AB506764.1 | - | 2 | 284 | ||
AK094237.1 | - | 2 | 1194 | |||
AK225393.1 | - | 2 | 332 | |||
AK299135.1 | - | 2 | 322 | |||
AK310028.1 | - | 2 | 293 | |||
AK311667.1 | - | 1 | 939 | |||
AY204547.1 | - | 2 | 284 | |||
AY750850.1 | - | 2 | 284 | |||
BC016929.1 | - | 2 | 338 | 4446575 | ||
BM552351.1 | - | 2 | 278 | |||
BX342160.2 | - | 2 | 308 | |||
CN483948.1 | - | 2 | 366 | |||
DT219772.1 | - | 2 | 278 | |||
L14848.1 | - | 2 | 323 |