Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PHLDB1 SIRNA Assays ›
Gene Symbol:
PHLDB1
Gene Name:
pleckstrin homology like domain family B member 1
Gene Aliases:
LL5A
Chromosome Location:
Chr.11: 118606296 - 118658038 on Build GRCh38
UniGene ID:
Hs.504062
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
217229
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PHLDB1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001144758.2
NP_001138230.1
12
2813
NM_001144759.2
NP_001138231.1
12
2813
NM_015157.3
NP_055972.1
13
3036
XM_005271468.3
XP_005271525.1
12
3582
XM_005271469.3
XP_005271526.1
12
3582
XM_005271470.3
XP_005271527.1
12
3582
XM_005271471.3
XP_005271528.1
12
3582
XM_005271472.3
XP_005271529.1
12
3582
XM_005271473.3
XP_005271530.1
12
3582
XM_005271474.4
XP_005271531.1
12
3582
XM_005277680.3
XP_005277737.1
13
3708
XM_006718796.3
XP_006718859.1
13
2897
XM_006718797.3
XP_006718860.1
13
3711
XM_006718798.3
XP_006718861.1
13
3711
XM_006718799.3
XP_006718862.1
13
3711
XM_006718800.3
XP_006718863.1
13
3711
XM_006718801.3
XP_006718864.1
13
3711
XM_006718802.3
XP_006718865.1
13
3711
XM_006718803.3
XP_006718866.1
13
3711
XM_011542703.2
XP_011541005.1
13
3711
XM_011542704.2
XP_011541006.1
13
3711
XM_011542705.2
XP_011541007.1
13
3711
XM_011542706.2
XP_011541008.1
13
3711
XM_011542707.2
XP_011541009.1
12
3582
XM_011542708.2
XP_011541010.1
13
2900
XM_011542709.2
XP_011541011.1
13
3210
XM_011542710.2
XP_011541012.1
13
3711
XM_011542711.2
XP_011541013.1
10
2617
XM_011542713.2
XP_011541015.1
10
2611
XM_011542715.2
XP_011541017.1
7
2089
XM_017017407.1
XP_016872896.1
12
3579
XM_017017408.1
XP_016872897.1
13
3711
XM_017017409.1
XP_016872898.1
13
3711
XM_017017410.1
XP_016872899.1
12
3582
GenBank mRNA
AB014538.2
-
12
2790
AB094090.1
-
13
3036
AB177859.1
-
7
1917
AK027510.1
-
4
442
AK074070.2
-
12
2762
AK074918.1
-
6
649
AK125783.1
-
14
2040
AL832373.1
-
12
2770
BC013031.2
-
6
799
BC098586.1
-
12
2790
30342198
BX648176.1
-
7
965
CR749783.1
-
13
2949
Back To Top
More Information
Gene Ontology Categories:
Function(s)
regulation of gastrulation
regulation of epithelial to mesenchymal transition
regulation of microtubule cytoskeleton organization
positive regulation of basement membrane assembly involved in embryonic body morphogenesis
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-fwm9x:80/10.239.164.221:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0