Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARHGAP27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282290.1 | NP_001269219.1 | 17 | 3360 | ||
NM_199282.2 | NP_954976.1 | 17 | 2768 | |||
XM_005257128.3 | XP_005257185.1 | 16 | 2817 | |||
XM_006721745.2 | XP_006721808.1 | 20 | 3742 | |||
XM_006721747.2 | XP_006721810.1 | 19 | 3676 | |||
XM_011524468.1 | XP_011522770.1 | 21 | 3841 | |||
XM_011524469.1 | XP_011522771.1 | 20 | 3775 | |||
XM_011524470.2 | XP_011522772.1 | 21 | 3831 | |||
XM_011524471.1 | XP_011522773.1 | 21 | 3670 | |||
XM_011524472.2 | XP_011522774.1 | 18 | 2982 | |||
XM_011524474.1 | XP_011522776.1 | 18 | 2852 | |||
XM_017024311.1 | XP_016879800.1 | 19 | 3775 | |||
XM_017024312.1 | XP_016879801.1 | 17 | 2881 | |||
GenBank mRNA | AK125535.1 | - | 17 | 2753 | ||
BC029634.1 | - | 1 | 423 |