Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KDM2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005366.1 | NP_001005366.1 | 5 | 739 | ||
NM_032590.4 | NP_115979.3 | 5 | 529 | |||
XM_005253955.3 | XP_005254012.1 | 5 | 739 | |||
XM_005253956.3 | XP_005254013.1 | 5 | 336 | |||
XM_011538867.2 | XP_011537169.1 | 5 | 1782 | |||
XM_011538868.2 | XP_011537170.1 | 5 | 1778 | |||
XM_011538869.2 | XP_011537171.1 | 5 | 1780 | |||
GenBank mRNA | AJ459424.1 | - | 5 | 501 | ||
AK097933.1 | - | 3 | 149 | |||
AK127328.1 | - | 5 | 739 | |||
AL133572.1 | - | 1 | 42 | |||
BC115379.1 | - | 4 | 389 | |||
BC115380.1 | - | 5 | 468 | 40029132 | ||
BX648928.1 | - | 5 | 531 |