Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sval3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003952.1 | NP_001003952.1 | 4 | 389 | ||
GenBank mRNA | AB098509.1 | - | 4 | 389 |