Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRC18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001006939.3 | NP_001006940.3 | 1 | 136 | ||
XM_006717867.2 | XP_006717930.1 | 3 | 613 | |||
XM_011539822.1 | XP_011538124.1 | 4 | 749 | |||
XM_011539823.1 | XP_011538125.1 | 3 | 616 | |||
XM_011539826.2 | XP_011538128.1 | 2 | 1903 | |||
XM_011539828.2 | XP_011538130.1 | 2 | 1442 | |||
XM_011539829.2 | XP_011538131.1 | 2 | 1445 | |||
XM_017016276.1 | XP_016871765.1 | 4 | 752 | |||
XM_017016277.1 | XP_016871766.1 | 2 | 1906 | |||
GenBank mRNA | AY358137.1 | - | 2 | 157 | ||
BC036722.1 | - | 1 | 147 | 5206652 |