Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Noa1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001006959.1 | NP_001006960.1 | 1 | 945 | ||
XM_006250845.2 | XP_006250907.1 | 1 | 1260 | |||
GenBank mRNA | BC083902.1 | - | 1 | 945 | 7133605 |