Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcgf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007000.1 | NP_001007001.1 | 4 | 336 | ||
GenBank mRNA | AY662670.1 | - | 4 | 336 |