Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CROCC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014675.4 | NP_055490.4 | 20 | 2954 | ||
XM_006711058.3 | XP_006711121.1 | 19 | 2745 | |||
XM_011542468.1 | XP_011540770.1 | 20 | 2931 | |||
XM_011542470.2 | XP_011540772.1 | 20 | 2931 | |||
XM_017002913.1 | XP_016858402.1 | 20 | 2931 | |||
XM_017002914.1 | XP_016858403.1 | 20 | 2931 | |||
GenBank mRNA | AB007914.2 | - | 18 | 2593 | ||
BC126911.1 | - | 17 | 2475 | |||
BC126912.1 | - | 17 | 2475 | |||
DQ139275.1 | - | 7 | 2311 |
Gene Symbol: CROCCP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK054737.1 | - | 1 | 1077 | ||
AK090414.1 | - | 11 | 4713 |