Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Emr4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007558.1 | NP_001007559.1 | 436 | |||
XM_008766767.1 | XP_008764989.1 | 424 | ||||
GenBank mRNA | AY686633.1 | - | 436 |
Set Membership: |
G-protein coupled receptor |