Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp58 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007575.2 | NP_001007576.1 | 3, 4 | 379 | ||
GenBank mRNA | AK041250.1 | - | 3, 4 | 315 | ||
AK162259.1 | - | 3, 4 | 329 | |||
AK167399.1 | - | 3, 4 | 321 | |||
DV071305.1 | - | 3, 4 | 407 |