Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Best3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007583.1 | NP_001007584.1 | 7 | 1310 | ||
GenBank mRNA | AY450426.1 | - | 7 | 1310 |