Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ubald1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007668.1 | NP_001007669.1 | 1 | 94 | ||
GenBank mRNA | BC079459.1 | - | 1 | 94 | 7121902 | |
FQ214502.1 | - | 1 | 131 | |||
FQ228825.1 | - | 1 | 122 |
Gene Symbol: LOC100910875 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003750772.3 | XP_003750820.1 | 792 |
Gene Symbol: Ubald2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_213532.7 | XP_213532.4 | 1 | 632 |