Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZFYVE16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105251.2 | NP_001098721.1 | 4 | 574 | ||
NM_001284236.1 | NP_001271165.1 | 4 | 500 | |||
NM_001284237.1 | NP_001271166.1 | 3 | 446 | |||
NM_014733.4 | NP_055548.3 | 3 | 446 | |||
XM_005248632.4 | XP_005248689.1 | 3 | 572 | |||
XM_011543753.2 | XP_011542055.1 | 4 | 471 | |||
XM_017010091.1 | XP_016865580.1 | 5 | 632 | |||
XM_017010092.1 | XP_016865581.1 | 4 | 618 | |||
XM_017010095.1 | XP_016865584.1 | 4 | 471 | |||
GenBank mRNA | AB002303.1 | - | 4 | 514 | ||
AF434817.1 | - | 2 | 266 | |||
BC030808.1 | - | 3 | 376 | 4701661 | ||
BX537424.1 | - | 3 | 398 | |||
BX956000.1 | - | 3 | 445 | |||
CR933621.1 | - | 4 | 449 | |||
DA216169.1 | - | 3 | 446 | |||
DR005686.1 | - | 4 | 441 |