Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cndp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007687.1 | NP_001007688.1 | 7 | 1027 | ||
XM_006255009.2 | XP_006255071.1 | 8 | 2749 | |||
XM_006255010.2 | XP_006255072.1 | 8 | 2750 | |||
XM_008772135.1 | XP_008770357.1 | 8 | 1049 | |||
GenBank mRNA | BC081877.1 | - | 7 | 1027 | 7107931 |