Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nfatc2ip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007692.1 | NP_001007693.1 | 8 | 1402 | ||
GenBank mRNA | BC079050.1 | - | 8 | 1402 | 7115323 |