Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trmt61a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007706.1 | NP_001007707.1 | 4 | 1815 | ||
XM_008764938.1 | XP_008763160.1 | 3 | 2301 | |||
GenBank mRNA | BC079198.1 | - | 4 | 1815 | 7131557 |
Gene Symbol: LOC103692719 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008764975.1 | XP_008763197.1 | 2301 | |||
XM_008764976.1 | XP_008763198.1 | 1814 |