Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TBC1D5 SIRNA Assays ›
Gene Symbol:
TBC1D5
Gene Name:
TBC1 domain family member 5
Gene Aliases:
-
Chromosome Location:
Chr.3: 17157162 - 17742739 on Build GRCh38
UniGene ID:
Hs.475629
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
21815
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TBC1D5
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001134381.1
NP_001127853.1
8
655
NM_014744.2
NP_055559.1
7
2056
XM_005265611.1
XP_005265668.1
7
626
XM_005265612.1
XP_005265669.1
6
561
XM_005265614.1
XP_005265671.1
7
626
XM_005265615.1
XP_005265672.1
6
556
XM_005265616.4
XP_005265673.1
5
485
XM_006713430.1
XP_006713493.1
7
676
XM_011534281.1
XP_011532583.1
9
937
XM_011534283.2
XP_011532585.1
8
715
XM_011534284.2
XP_011532586.1
8
760
XM_011534286.1
XP_011532588.1
7
692
XM_011534287.1
XP_011532589.1
7
674
XM_017007552.1
XP_016863041.1
10
1038
XM_017007553.1
XP_016863042.1
10
1068
XM_017007554.1
XP_016863043.1
15
1681
XM_017007555.1
XP_016863044.1
9
920
XM_017007556.1
XP_016863045.1
8
900
XM_017007557.1
XP_016863046.1
6
520
XM_017007558.1
XP_016863047.1
6
550
XM_017007559.1
XP_016863048.1
8
900
XM_017007560.1
XP_016863049.1
8
730
XM_017007561.1
XP_016863050.1
6
561
XM_017007562.1
XP_016863051.1
7
674
XM_017007563.1
XP_016863052.1
5
480
XM_017007564.1
XP_016863053.1
6
556
XM_017007565.1
XP_016863054.1
7
854
XM_017007566.1
XP_016863055.1
7
643
XM_017007567.1
XP_016863056.1
5
484
XM_017007568.1
XP_016863057.1
6
520
GenBank mRNA
AB449886.1
-
5
391
AK309374.1
-
3
316
AK310539.1
-
8
655
BC013145.1
-
6
568
4510019
D86965.1
-
7
2055
Back To Top
More Information
Panther Classification:
Molecular Function -
GTPase-activating protein
Gene Ontology Categories:
Function(s)
Process(es)
positive regulation of receptor internalization
intracellular protein transport
positive regulation of autophagy
regulation of vesicle fusion
retrograde transport, endosome to Golgi
response to starvation
activation of GTPase activity
mitophagy in response to mitochondrial depolarization
regulation of cilium assembly
GTPase activator activity
protein binding
Rab GTPase binding
AP-2 adaptor complex binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-x5whk:80/10.239.163.84:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0