Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Emp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007721.1 | NP_001007722.1 | 3, 4 | 343 | ||
GenBank mRNA | BC081865.1 | - | 3, 4 | 343 | 7096470 |