Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nt5c3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007723.2 | NP_001007724.2 | 7 | 609 | ||
XM_008768083.1 | XP_008766305.1 | 5 | 427 | |||
XM_008768084.1 | XP_008766306.1 | 5 | 427 | |||
GenBank mRNA | BC078963.1 | - | 7 | 609 | 7111222 |