Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Glyr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007800.1 | NP_001007801.1 | 6 | 591 | ||
XM_006245803.2 | XP_006245865.1 | 6 | 660 | |||
XM_006245804.2 | XP_006245866.1 | 5 | 420 | |||
GenBank mRNA | BC085931.1 | - | 6 | 602 | 7111787 |