Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Daam2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008231.2 | NP_001008232.2 | 6 | 713 | ||
GenBank mRNA | AK122266.1 | - | 6 | 713 | ||
AY426536.1 | - | 6 | 742 | |||
BC050043.1 | - | 6 | 713 |