Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008277.1 | NP_001008278.1 | 21 | 2249 | ||
GenBank mRNA | BC085837.1 | - | 21 | 2249 | 7135552 |
Set Membership: |
G-protein coupled receptor |