Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Usp14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008301.1 | NP_001008302.1 | 9 | 836 | ||
XM_006254396.2 | XP_006254458.1 | 9 | 1030 | |||
XM_006254398.2 | XP_006254460.1 | 8 | 824 | |||
GenBank mRNA | BC085947.1 | - | 9 | 836 | 7130090 |