Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLPP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003713.4 | NP_003704.3 | 6 | 1986 | ||
GenBank mRNA | AF480883.1 | - | 7 | 1798 | ||
BM719257.1 | - | 1 | 93 | |||
BU608248.1 | - | 1 | 471 |